Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015295.2 | NP_056110.2 | 26 | 3602 | ||
XM_011525642.1 | XP_011523944.1 | 26 | 3602 | |||
XM_011525643.2 | XP_011523945.1 | 26 | 3602 | |||
XM_011525644.2 | XP_011523946.1 | 25 | 3510 | |||
XM_017025684.1 | XP_016881173.1 | 27 | 3780 | |||
GenBank mRNA | AK126324.1 | - | 11 | 1233 | ||
CR627458.1 | - | 15 | 1826 | |||
DA283721.1 | - | 4 | 554 | |||
DA675822.1 | - | 3 | 457 |