Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMCHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015295.2 | NP_056110.2 | 31 | 4131 | ||
XM_011525642.1 | XP_011523944.1 | 31 | 4131 | |||
XM_011525643.2 | XP_011523945.1 | 31 | 4131 | |||
XM_011525644.2 | XP_011523946.1 | 30 | 4039 | |||
XM_017025684.1 | XP_016881173.1 | 32 | 4309 | |||
GenBank mRNA | AB014550.1 | - | 5 | 473 | ||
AK126324.1 | - | 16 | 1762 | |||
AL080138.1 | - | 2 | 111 | |||
CR627458.1 | - | 20 | 2355 |