Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166133.1 | NP_001159605.1 | 11 | 1527 | ||
NM_025074.6 | NP_079350.5 | 11 | 1527 | |||
XM_006714316.2 | XP_006714379.1 | 11 | 1774 | |||
GenBank mRNA | AK027833.1 | - | 6 | 512 | ||
BC020192.1 | - | 3 | 307 | |||
BC052281.1 | - | 6 | 614 | |||
BC064487.1 | - | 6 | 614 | |||
BC094853.1 | - | 3 | 307 | |||
BC131820.1 | - | 11 | 1117 |