Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282352.1 | NP_001269281.1 | 3 | 378 | ||
NM_030957.3 | NP_112219.3 | 16 | 2178 | |||
XM_006722917.3 | XP_006722980.1 | 15 | 2024 | |||
XM_017027338.1 | XP_016882827.1 | 15 | 2034 | |||
XM_017027339.1 | XP_016882828.1 | 6 | 658 | |||
XM_017027340.1 | XP_016882829.1 | 5 | 518 | |||
GenBank mRNA | AB209515.1 | - | 15 | 2014 | ||
AF163762.1 | - | 13 | 1794 | |||
AK131414.1 | - | 3 | 378 |