Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152641.2 | NP_689854.2 | 15 | 2108 | ||
XM_006719272.3 | XP_006719335.1 | 15 | 2262 | |||
XM_011538025.1 | XP_011536327.1 | 9 | 1215 | |||
GenBank mRNA | AL832200.1 | - | 11 | 1661 | ||
AY727870.2 | - | 15 | 2108 | |||
BX648152.1 | - | 9 | 1399 | |||
BX649133.1 | - | 7 | 954 | |||
CR749833.1 | - | 14 | 2027 | |||
DQ096628.1 | - | 15 | 2108 |