Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032504.1 | NP_115893.1 | 50 | 7565 | ||
NM_182587.3 | NP_872393.3 | 50 | 7550 | |||
XM_005246476.1 | XP_005246533.1 | 51 | 7760 | |||
XM_011511004.1 | XP_011509306.1 | 51 | 7805 | |||
XM_011511005.1 | XP_011509307.1 | 51 | 7802 | |||
XM_011511006.1 | XP_011509308.1 | 51 | 7799 | |||
XM_011511007.1 | XP_011509309.1 | 51 | 7769 | |||
XM_011511008.1 | XP_011509310.1 | 51 | 7805 | |||
XM_011511010.2 | XP_011509312.1 | 51 | 7805 | |||
XM_017003884.1 | XP_016859373.1 | 51 | 7790 | |||
XM_017003885.1 | XP_016859374.1 | 51 | 7763 | |||
XM_017003886.1 | XP_016859375.1 | 51 | 7805 | |||
XM_017003887.1 | XP_016859376.1 | 50 | 7601 | |||
XM_017003888.1 | XP_016859377.1 | 50 | 7559 | |||
XM_017003889.1 | XP_016859378.1 | 50 | 7556 | |||
XM_017003890.1 | XP_016859379.1 | 50 | 7544 | |||
XM_017003891.1 | XP_016859380.1 | 50 | 7824 | |||
XM_017003892.1 | XP_016859381.1 | 38 | 6608 | |||
XM_017003894.1 | XP_016859383.1 | 29 | 4036 | |||
GenBank mRNA | AB058746.2 | - | 36 | 5010 | ||
AK027583.1 | - | 1 | 63 |