Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEX3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174118.1 | NP_001167589.1 | 2 | 669 | ||
NM_203304.3 | NP_976049.3 | 2 | 669 | |||
XM_017026811.1 | XP_016882300.1 | 2 | 469 | |||
GenBank mRNA | AB107353.1 | - | 2 | 262 | ||
AF458084.1 | - | 2 | 141 | |||
AY950680.1 | - | 2 | 669 | |||
BC113384.1 | - | 2 | 140 | |||
BC113741.1 | - | 2 | 140 | |||
DQ225272.1 | - | 3 | 339 | |||
DQ225273.1 | - | 3 | 300 |