Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000517.4 | NP_000508.1 | 3 | 584 |
Gene Symbol: HBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF147332.1 | - | 3 | 536 | ||
BC008572.1 | - | 3 | 541 | 4177243 | ||
V00493.1 | - | 3 | 555 |