Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016219.4 | NP_057303.2 | 3 | 482 | ||
NR_045720.1 | 3 | 482 | ||||
NR_045721.1 | 4 | 613 | ||||
XM_006716945.3 | XP_006717008.1 | 3 | 547 | |||
XM_017014239.1 | XP_016869728.1 | 4 | 459 | |||
GenBank mRNA | AF145732.1 | - | 3 | 399 | ||
AF148509.1 | - | 3 | 459 | |||
AK298480.1 | - | 4 | 459 | |||
AK299043.1 | - | 3 | 329 | |||
AK309808.1 | - | 4 | 459 | |||
AK315797.1 | - | 3 | 424 | |||
AY358465.1 | - | 3 | 431 | |||
BC002953.1 | - | 3 | 415 | 3533651 | ||
BC006079.1 | - | 3 | 424 | 3959196 | ||
BE731944.1 | - | 508 | ||||
CR749534.1 | - | 4 | 455 |