Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016219.4 | NP_057303.2 | 13 | 2000 | ||
NR_045720.1 | 13 | 1975 | ||||
NR_045721.1 | 14 | 2131 | ||||
XM_006716945.3 | XP_006717008.1 | 12 | 2790 | |||
XM_017014239.1 | XP_016869728.1 | 13 | 2702 | |||
GenBank mRNA | AF145732.1 | - | 13 | 1917 | ||
AF148509.1 | - | 13 | 1977 | |||
AK074559.1 | - | 5 | 564 | |||
AK074699.1 | - | 11 | 1575 | |||
AK094878.1 | - | 5 | 2006 | |||
AK126926.1 | - | 6 | 4034 | |||
AK127005.1 | - | 6 | 3099 | |||
AK128033.1 | - | 5 | 4285 | |||
AK128257.1 | - | 6 | 3177 | |||
AK128341.1 | - | 6 | 3847 | |||
AK130143.1 | - | 8 | 1144 | |||
AK298480.1 | - | 14 | 1935 | |||
AK315797.1 | - | 13 | 1917 | |||
AL110221.1 | - | 5 | 1447 | |||
AY358465.1 | - | 13 | 1949 | |||
BC002953.1 | - | 13 | 1933 | 3533651 | ||
BC006079.1 | - | 13 | 1942 | 3959196 | ||
CR749534.1 | - | 19 | 6483 |