Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACVR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020328.3 | NP_064733.3 | 5 | 1002 | ||
XM_011538966.2 | XP_011537268.1 | 5 | 980 | |||
XM_017020199.1 | XP_016875688.1 | 5 | 1700 | |||
XM_017020200.1 | XP_016875689.1 | 6 | 1095 | |||
GenBank mRNA | AK299120.1 | - | 5 | 965 |