Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278616.1 | NP_001265545.1 | 24 | 3283 | ||
NM_001278617.1 | NP_001265546.1 | 24 | 3172 | |||
NM_004336.4 | NP_004327.1 | 25 | 3343 | |||
GenBank mRNA | AF011387.1 | - | 17 | 2408 | ||
AF043294.2 | - | 25 | 3263 | |||
AF046078.1 | - | 25 | 3280 | |||
AF047471.1 | - | 25 | 3266 | |||
AF053305.1 | - | 25 | 3294 | |||
AK225317.1 | - | 24 | 3128 | |||
AK291221.1 | - | 25 | 3296 | |||
AK302418.1 | - | 24 | 3239 | |||
BC028201.1 | - | 25 | 3264 | 5205385 |