Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL9A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001853.3 | NP_001844.3 | 32 | 2047 | ||
GenBank mRNA | AK075240.1 | - | 8 | 1771 | ||
BC004412.1 | - | 4 | 555 | |||
BC007574.1 | - | 25 | 1677 | |||
BC011705.2 | - | 32 | 2047 | 3830925 | ||
L41162.1 | - | 32 | 2044 |