Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF3R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000760.3 | NP_000751.1 | 17 | 3345 | ||
NM_156039.3 | NP_724781.1 | 17 | 3426 | |||
NM_172313.2 | NP_758519.1 | 18 | 2925 | |||
XM_005270493.1 | XP_005270550.1 | 17 | 3342 | |||
XM_011540748.2 | XP_011539050.1 | 16 | 2951 | |||
XM_011540749.1 | XP_011539051.1 | 17 | 3423 | |||
XM_011540750.1 | XP_011539052.1 | 12 | 2256 | |||
XM_017000370.1 | XP_016855859.1 | 16 | 2988 | |||
GenBank mRNA | AB209656.1 | - | 9 | 6011 | ||
BC053585.1 | - | 17 | 2938 | 5217305 | ||
M59818.1 | - | 17 | 2930 | |||
M59819.1 | - | 17 | 2842 | |||
M59820.1 | - | 17 | 3011 | |||
X55720.1 | - | 18 | 2505 |