Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080449.2 | NP_001073918.2 | 6 | 1021 | ||
NR_102264.1 | 7 | 1001 | ||||
XM_006717680.3 | XP_006717743.1 | 7 | 1401 | |||
XM_011539417.1 | XP_011537719.1 | 1 | 63 | |||
XM_017015799.1 | XP_016871288.1 | 3 | 410 | |||
GenBank mRNA | AK308333.1 | - | 6 | 1021 | ||
AK308358.1 | - | 4 | 662 | |||
BC063664.1 | - | 6 | 934 | |||
BC111740.1 | - | 7 | 1001 | 40038149 | ||
D42046.1 | - | 6 | 963 |