Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000860.5 | NP_000851.2 | 6 | 1025 | ||
NM_001256301.1 | NP_001243230.1 | 6 | 755 | |||
NM_001256306.1 | NP_001243235.1 | 4 | 821 | |||
NM_001256307.1 | NP_001243236.1 | 5 | 918 | |||
GenBank mRNA | AK058013.1 | - | 6 | 614 | ||
AK296642.1 | - | 6 | 755 | |||
AK300125.1 | - | 4 | 411 | |||
AK300524.1 | - | 5 | 661 | |||
AK314624.1 | - | 6 | 614 | |||
BC018986.2 | - | 6 | 587 | 3638799 | ||
J05594.1 | - | 6 | 595 | |||
L76465.1 | - | 6 | 595 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DC424637.1 | - | 7 | 663 | Chr. 4 - 174493163 - 174523081 [-] on Build GRCh38 |