Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000860.5 | NP_000851.2 | 7 | 1203 | ||
NM_001145816.2 | NP_001139288.1 | 6 | 1039 | |||
NM_001256301.1 | NP_001243230.1 | 7 | 933 | |||
NM_001256305.1 | NP_001243234.1 | 5 | 962 | |||
NM_001256306.1 | NP_001243235.1 | 5 | 999 | |||
NM_001256307.1 | NP_001243236.1 | 6 | 1096 | |||
GenBank mRNA | AK058013.1 | - | 7 | 792 | ||
AK296642.1 | - | 7 | 933 | |||
AK300125.1 | - | 5 | 589 | |||
AK300524.1 | - | 6 | 839 | |||
AK300940.1 | - | 6 | 628 | |||
AK314624.1 | - | 7 | 792 | |||
BC018986.2 | - | 7 | 765 | 3638799 | ||
J05594.1 | - | 7 | 773 | |||
L76465.1 | - | 7 | 773 | |||
U63296.1 | - | 5 | 515 | |||
X82460.1 | - | 6 | 592 |