Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_078471.3 | NP_510880.2 | 18 | 3174 | ||
NM_203318.1 | NP_976063.1 | 18 | 3174 | |||
GenBank mRNA | AB177858.1 | - | 17 | 2993 | ||
AB177860.1 | - | 18 | 2126 | |||
AK302505.1 | - | 17 | 1927 | |||
AK303522.1 | - | 17 | 2064 | |||
AY703984.1 | - | 18 | 2126 | |||
BC039612.1 | - | 18 | 3278 | 6045482 | ||
CR933614.1 | - | 18 | 3149 | |||
D86970.2 | - | 18 | 3197 |