Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008541.1 | NP_001008541.1 | 4 | 464 | ||
NM_005962.4 | NP_005953.4 | 5 | 689 | |||
NM_130439.3 | NP_569157.2 | 5 | 887 | |||
GenBank mRNA | AK307090.1 | - | 6 | 575 | ||
AK309718.1 | - | 5 | 502 | |||
AL538051.3 | - | 5 | 700 | |||
BC012907.1 | - | 4 | 464 | 3882557 | ||
BC035128.1 | - | 5 | 893 | 5263647 | ||
BG700036.1 | - | 5 | 673 | |||
BM927870.1 | - | 5 | 672 | |||
BT007069.1 | - | 3 | 344 | |||
BX417051.2 | - | 5 | 685 | |||
CR536576.1 | - | 5 | 482 | |||
D63940.1 | - | 4 | 374 | |||
KU178207.1 | - | 3 | 344 | |||
KU178208.1 | - | 4 | 390 | |||
KU178209.1 | - | 4 | 374 |