Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002508.2 | NP_002499.2 | 20 | 3827 | ||
XM_011544195.2 | XP_011542497.1 | 19 | 3698 | |||
GenBank mRNA | AB209448.1 | - | 20 | 3709 | ||
BC045606.1 | - | 17 | 3338 | 5271590 | ||
DA269707.1 | - | 5 | 533 | |||
M27445.1 | - | 12 | 1727 | |||
M30269.1 | - | 20 | 3815 |