Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291516.1 | NP_001278445.1 | 5 | 791 | ||
NR_111982.1 | 5 | 1108 | ||||
XM_011518679.2 | XP_011516981.1 | 5 | 1114 | |||
XM_011518681.2 | XP_011516983.1 | 5 | 1113 | |||
XM_011518683.2 | XP_011516985.1 | 5 | 1113 | |||
XM_011518684.2 | XP_011516986.1 | 5 | 1113 | |||
GenBank mRNA | BC031926.1 | - | 5 | 791 | ||
BC042107.1 | - | 5 | 1084 | 5170879 |