Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166292.1 | NP_001159764.1 | 3, 4 | 477 | ||
NM_003738.4 | NP_003729.3 | 3, 4 | 477 | |||
GenBank mRNA | AF087651.1 | - | 3, 4 | 465 | ||
AF091501.1 | - | 3, 4 | 465 | |||
AF119569.1 | - | 3, 4 | 477 | |||
AY358555.1 | - | 3, 4 | 465 | |||
AY359016.1 | - | 3, 4 | 469 | |||
BC143712.1 | - | 3, 4 | 533 |