Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003863.3 | NP_003854.1 | 4 | 1270 | ||
GenBank mRNA | AF061729.1 | - | 5 | 634 | ||
BC015233.1 | - | 4 | 1270 | 3901944 | ||
BC015374.1 | - | 2 | 1390 | 4181887 | ||
BC107863.1 | - | 4 | 644 | 6450798 |