Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRIPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014171.5 | NP_054890.1 | 3 | 214 | ||
GenBank mRNA | AF161488.1 | - | 3 | 190 | ||
AK291298.1 | - | 3 | 198 | |||
BC006980.1 | - | 3 | 171 | 3826521 | ||
BC018653.1 | - | 3 | 178 | 4080544 | ||
BI857531.1 | - | 273 | ||||
BU603377.1 | - | 217 |