Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185075.1 | NP_001172004.1 | 8 | 895 | ||
NM_001185076.1 | NP_001172005.1 | 8 | 895 | |||
NM_001185081.1 | NP_001172010.1 | 8 | 895 | |||
NM_001185082.1 | NP_001172011.1 | 8 | 895 | |||
NM_002024.5 | NP_002015.1 | 8 | 895 | |||
NR_033699.1 | 8 | 895 | ||||
NR_033700.1 | 8 | 895 | ||||
GenBank mRNA | AB209188.1 | - | 4 | 1385 | ||
AK302707.1 | - | 2 | 79 | |||
BC038998.1 | - | 8 | 860 | |||
BC086957.1 | - | 8 | 925 | 30347992 | ||
BQ417290.1 | - | 4 | 343 | |||
CK825936.1 | - | 4 | 291 | |||
KC774027.1 | - | 8 | 728 | |||
KC774028.1 | - | 8 | 728 | |||
KC774029.1 | - | 8 | 728 | |||
KF856234.1 | - | 8 | 728 | |||
KJ534836.1 | - | 8 | 666 | |||
KJ534837.1 | - | 8 | 666 | |||
KJ534838.1 | - | 8 | 666 | |||
KM091457.1 | - | 8 | 728 | |||
KM091458.1 | - | 8 | 728 | |||
KM091459.1 | - | 8 | 728 | |||
M67468.1 | - | 8 | 861 | |||
S65791.1 | - | 8 | 885 | |||
X69962.1 | - | 8 | 885 |