Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006063.2 | NP_006054.2 | 1 | 784 | ||
GenBank mRNA | AF056929.1 | - | 1 | 754 | ||
AF333387.1 | - | 1 | 784 | |||
AK316571.1 | - | 1 | 781 | |||
AY364252.1 | - | 1 | 194 | |||
BC006534.2 | - | 1 | 762 | 2959774 |