Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282716.1 | NP_001269645.1 | 26 | 2974 | ||
NM_001282717.1 | NP_001269646.1 | 26 | 3055 | |||
NM_001282718.1 | NP_001269647.1 | 24 | 2848 | |||
NM_012447.3 | NP_036579.2 | 26 | 3055 | |||
XM_011515742.1 | XP_011514044.1 | 26 | 3022 | |||
XM_017011683.1 | XP_016867172.1 | 26 | 2902 | |||
XM_017011684.1 | XP_016867173.1 | 26 | 3022 | |||
XM_017011685.1 | XP_016867174.1 | 26 | 3135 | |||
XM_017011686.1 | XP_016867175.1 | 26 | 2902 | |||
XM_017011687.1 | XP_016867176.1 | 24 | 2728 | |||
GenBank mRNA | AJ007798.2 | - | 26 | 2975 | ||
AK292394.1 | - | 26 | 2882 | |||
AK302587.1 | - | 23 | 2466 | |||
AK302693.1 | - | 24 | 2671 | |||
BC140932.1 | - | 26 | 2814 | 9056744 | ||
BC146806.1 | - | 26 | 2880 | 8860345 |