Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014049.4 | NP_054768.2 | 13, 14 | 1559 | ||
NR_033426.1 | 13, 14 | 1735 | ||||
XM_017006264.1 | XP_016861753.1 | 13, 14 | 1723 | |||
XM_017006265.1 | XP_016861754.1 | 7, 8 | 709 | |||
GenBank mRNA | AF078854.1 | - | 1, 2 | 94 | ||
AF327351.1 | - | 13, 14 | 1389 | |||
AK022568.1 | - | 12, 13 | 1485 | |||
AK024012.1 | - | 13, 14 | 1403 | |||
AK027186.1 | - | 3, 4 | 381 | |||
AK130311.1 | - | 13, 14 | 1430 | |||
AK297952.1 | - | 13, 14 | 1603 | |||
BC001817.2 | - | 6, 7 | 523 | |||
BC007970.2 | - | 13, 14 | 1436 | 4304209 | ||
BC013354.2 | - | 13, 14 | 1413 | 3935925 |