Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSDMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042471.1 | NP_001035936.1 | 6, 7 | 812 | ||
NM_001165958.1 | NP_001159430.1 | 7, 8 | 851 | |||
XM_011525004.1 | XP_011523306.1 | 7, 8 | 2259 | |||
XM_011525006.1 | XP_011523308.1 | 7, 8 | 2250 | |||
XM_011525007.2 | XP_011523309.1 | 7, 8 | 1581 | |||
XM_011525009.1 | XP_011523311.1 | 7, 8 | 1939 | |||
XM_011525011.1 | XP_011523313.1 | 7, 8 | 1590 | |||
XM_011525012.1 | XP_011523314.1 | 7, 8 | 1930 | |||
XM_011525013.1 | XP_011523315.1 | 8, 9 | 1648 | |||
XM_011525015.1 | XP_011523317.1 | 8, 9 | 1639 | |||
XM_017024848.1 | XP_016880337.1 | 7, 8 | 2582 | |||
XM_017024849.1 | XP_016880338.1 | 8, 9 | 2291 | |||
XM_017024850.1 | XP_016880339.1 | 6, 7 | 2781 | |||
XM_017024852.1 | XP_016880341.1 | 5, 6 | 2742 | |||
GenBank mRNA | AK000409.1 | - | 6, 7 | 752 | ||
BP366854.1 | - | 4, 5 | 503 | |||
BX538068.1 | - | 6, 7 | 515 | |||
CA488357.1 | - | 2, 3 | 232 | |||
EU526753.1 | - | 5, 6 | 681 | |||
EU526755.1 | - | 4, 5 | 596 | |||
EU526756.1 | - | 3, 4 | 427 |