Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPV4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177428.1 | NP_001170899.1 | 6 | 1313 | ||
NM_001177431.1 | NP_001170902.1 | 8 | 1321 | |||
NM_001177433.1 | NP_001170904.1 | 5 | 1133 | |||
NM_021625.4 | NP_067638.3 | 8 | 1512 | |||
NM_147204.2 | NP_671737.1 | 6 | 1274 | |||
XM_005253918.1 | XP_005253975.1 | 8 | 1872 | |||
XM_011538630.2 | XP_011536932.2 | 8 | 2615 | |||
XM_011538631.2 | XP_011536933.2 | 7 | 2475 | |||
XM_011538632.2 | XP_011536934.2 | 7 | 2436 | |||
XM_011538633.2 | XP_011536935.2 | 6 | 2295 | |||
XM_011538634.2 | XP_011536936.2 | 8 | 2617 | |||
XM_011538635.2 | XP_011536937.1 | 8 | 2619 | |||
XM_011538636.2 | XP_011536938.1 | 8 | 2620 | |||
XM_017019774.1 | XP_016875263.1 | 8 | 2245 | |||
GenBank mRNA | AB032427.1 | - | 7 | 1518 | ||
AB073669.1 | - | 6 | 1243 | |||
AB100308.1 | - | 7 | 1423 | |||
AF258465.1 | - | 7 | 1423 | |||
AF263523.1 | - | 7 | 1423 | |||
AF279673.1 | - | 7 | 1219 | |||
AJ296305.1 | - | 8 | 1512 | |||
AL834192.1 | - | 8 | 1483 | |||
BC117426.1 | - | 8 | 1506 | 40125977 | ||
BC143307.1 | - | 8 | 1506 | |||
BC143315.1 | - | 8 | 1506 | 9051820 | ||
DQ059644.1 | - | 6 | 1282 | |||
DQ059645.1 | - | 8 | 1321 | |||
DQ059646.1 | - | 5 | 1102 |