Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281520.1 | NP_001268449.1 | 13 | 1900 | ||
NM_001281521.1 | NP_001268450.1 | 13 | 1966 | |||
NM_001281522.1 | NP_001268451.1 | 12 | 1705 | |||
NM_032485.5 | NP_115874.3 | 13 | 1846 | |||
NM_182802.2 | NP_877954.1 | 13 | 1852 | |||
XM_011529387.2 | XP_011527689.1 | 13 | 1809 | |||
XM_017028105.1 | XP_016883594.1 | 13 | 1995 | |||
XM_017028106.1 | XP_016883595.1 | 13 | 1812 | |||
XM_017028107.1 | XP_016883596.1 | 6 | 2085 | |||
GenBank mRNA | AJ439063.1 | - | 13 | 1846 | ||
AK027644.1 | - | 7 | 885 | |||
AK314654.1 | - | 13 | 1528 | |||
AY158211.1 | - | 13 | 1551 | |||
BC005170.1 | - | 13 | 1551 | |||
BC008830.2 | - | 10 | 1156 | |||
BC080656.1 | - | 13 | 1551 | 4841271 | ||
BC101054.1 | - | 12 | 1506 | 40008094 | ||
BC101055.1 | - | 13 | 1767 | 40008095 | ||
BC101056.1 | - | 13 | 1647 | 40008096 | ||
BC101057.1 | - | 13 | 1701 | 40008097 | ||
KU178803.1 | - | 7 | 758 | |||
KU178804.1 | - | 8 | 849 | |||
KU178805.1 | - | 6 | 617 |