Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288781.1 | NP_001275710.1 | 13, 14 | 1478 | ||
NM_001288782.1 | NP_001275711.1 | 11, 12 | 1398 | |||
NM_001288783.1 | NP_001275712.1 | 12, 13 | 1370 | |||
NM_144596.3 | NP_653197.2 | 13, 14 | 1430 | |||
NM_198309.3 | NP_938051.1 | 12, 13 | 1400 | |||
NM_198310.3 | NP_938052.1 | 11, 12 | 1310 | |||
XM_006720035.1 | XP_006720098.1 | 11, 12 | 1682 | |||
XM_006720037.2 | XP_006720100.1 | 10, 11 | 1592 | |||
XM_011536432.1 | XP_011534734.1 | 12, 13 | 1760 | |||
XM_011536433.1 | XP_011534735.1 | 12, 13 | 1760 | |||
XM_011536434.1 | XP_011534736.1 | 11, 12 | 1670 | |||
XM_011536435.2 | XP_011534737.1 | 11, 12 | 1146 | |||
XM_017020977.1 | XP_016876466.1 | 11, 12 | 1682 | |||
XM_017020978.1 | XP_016876467.1 | 10, 11 | 1592 | |||
GenBank mRNA | AK093891.1 | - | 11, 12 | 1248 | ||
AK124675.1 | - | 12, 13 | 1280 | |||
AK315772.1 | - | 11, 12 | 1285 | |||
AL833901.1 | - | 10, 11 | 1134 | |||
AY366523.1 | - | 11, 12 | 1256 | |||
AY366524.1 | - | 10, 11 | 1166 | |||
AY373972.1 | - | 11, 12 | 1256 | |||
BC001563.1 | - | 7, 8 | 748 | |||
BC026351.1 | - | 12, 13 | 1334 | 4799330 | ||
BC095433.1 | - | 11, 12 | 1238 | 30368216 | ||
BX161472.1 | - | 10, 11 | 1238 | |||
BX247959.1 | - | 11, 12 | 1347 | |||
BX248071.1 | - | 3, 4 | 487 | |||
BX248248.1 | - | 8, 9 | 842 | |||
KU178849.1 | - | 12, 13 | 1282 | |||
KU178850.1 | - | 11, 12 | 1204 | |||
KU178851.1 | - | 10, 11 | 1114 | |||
KU178852.1 | - | 6, 7 | 724 | |||
KU178853.1 | - | 5, 6 | 549 |