Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT8L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178557.3 | NP_848652.2 | 3 | 641 | ||
GenBank mRNA | AK094797.1 | - | 3 | 293 | ||
BC093906.1 | - | 2 | 174 | 7939751 | ||
BC093908.1 | - | 2 | 175 | 7939753 | ||
BC103748.1 | - | 3 | 309 | 6195512 |