Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185076.1 | NP_001172005.1 | 13 | 1560 | ||
NM_001185082.1 | NP_001172011.1 | 13 | 1560 | |||
NM_002024.5 | NP_002015.1 | 14 | 1623 | |||
GenBank mRNA | AB209188.1 | - | 9 | 2050 | ||
AK294104.1 | - | 3 | 343 | |||
AK302707.1 | - | 8 | 884 | |||
BC086957.1 | - | 13 | 1590 | 30347992 | ||
CN315330.1 | - | 4 | 434 | |||
CX871385.1 | - | 2 | 198 | |||
KC774027.1 | - | 13 | 1393 | |||
KC774029.1 | - | 12 | 1258 | |||
KF856234.1 | - | 12 | 1258 | |||
KJ534836.1 | - | 13 | 1331 | |||
KJ534837.1 | - | 14 | 1394 | |||
KJ534838.1 | - | 13 | 1331 | |||
KM091457.1 | - | 15 | 1502 | |||
KM091458.1 | - | 14 | 1439 | |||
KM091459.1 | - | 15 | 1502 | |||
M67468.1 | - | 13 | 1520 | |||
S65791.1 | - | 14 | 1613 | |||
X69962.1 | - | 14 | 1613 |