Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033024.1 | NP_001028196.1 | 3 | 489 | ||
NM_001257990.1 | NP_001244919.1 | 3 | 641 | |||
NM_012179.3 | NP_036311.3 | 3 | 884 | |||
XM_011530106.1 | XP_011528408.1 | 2 | 627 | |||
GenBank mRNA | AF233225.1 | - | 3 | 604 | ||
AK226087.1 | - | 3 | 799 | |||
AK291972.1 | - | 3 | 624 | |||
AK297841.1 | - | 3 | 444 | |||
AK301716.1 | - | 3 | 569 | |||
AL050254.1 | - | 3 | 806 | |||
BC008361.1 | - | 3 | 881 | 3611049 | ||
BI821903.1 | - | 3 | 489 | |||
BX362053.2 | - | 3 | 856 | |||
BX648151.1 | - | 3 | 437 | |||
BX648220.1 | - | 3 | 434 | |||
CR456491.1 | - | 3 | 719 |