Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POMT1 SIRNA Assays ›
Gene Symbol:
POMT1
Gene Name:
protein O-mannosyltransferase 1
Gene Aliases:
LGMD2K, MDDGA1, MDDGB1, MDDGC1, RT
Chromosome Location:
Chr.9: 131502895 - 131523806 on Build GRCh38
UniGene ID:
Hs.522449
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s531709
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POMT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077365.1
NP_001070833.1
13
1422
NM_001077366.1
NP_001070834.1
12
1270
NM_001136113.1
NP_001129585.1
13
1662
NM_001136114.1
NP_001129586.1
11
1163
NM_007171.3
NP_009102.3
13
1488
XM_005272156.1
XP_005272213.1
13
1614
XM_005272158.1
XP_005272215.1
12
1343
XM_005272159.1
XP_005272216.1
11
1233
XM_005272162.2
XP_005272219.1
12
1664
XM_006716932.1
XP_006716995.1
12
1266
XM_011518140.1
XP_011516442.1
12
1341
XM_011518141.1
XP_011516443.1
12
1275
XM_011518142.1
XP_011516444.1
11
1143
XM_011518143.1
XP_011516445.1
11
1173
XM_011518145.1
XP_011516447.1
10
1009
XM_017014203.1
XP_016869692.1
13
1936
XM_017014204.1
XP_016869693.1
12
1210
XM_017014205.1
XP_016869694.1
13
1415
GenBank mRNA
AB209430.1
-
7
3118
AF095136.1
-
13
1465
AK000475.1
-
2
77
AK074874.1
-
13
1380
AK074888.1
-
12
1254
AK094558.1
-
3
1775
AK293529.1
-
11
1075
AK295458.1
-
7
701
AK295561.1
-
11
1167
AK299646.1
-
10
974
AK300393.1
-
12
1290
AK300511.1
-
5
549
AK315218.1
-
12
1625
BC022877.2
-
13
1316
4826806
BC065268.1
-
13
1607
6155142
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
cell wall mannoprotein biosynthetic process
carbohydrate metabolic process
protein O-linked glycosylation
multicellular organism development
extracellular matrix organization
protein O-linked mannosylation
chain elongation of O-linked mannose residue
regulation of endoplasmic reticulum unfolded protein response
mannosyltransferase activity
dolichyl-phosphate-mannose-protein mannosyltransferase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline