Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMS1P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024495.2 | 5 | 615 | |||
GenBank mRNA | BC051298.1 | - | 4 | 581 | ||
CN417542.1 | - | 4 | 576 | |||
CN417548.1 | - | 3 | 262 |
Gene Symbol: BMS1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003611.2 | 5 | 733 | |||
GenBank mRNA | AK307333.1 | - | 5 | 592 | ||
AL832069.1 | - | 5 | 620 | |||
BC017177.2 | - | 4 | 581 | |||
BC131568.1 | - | 4 | 1387 |