Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198542.1 | NP_001185471.1 | 2, 3 | 245 | ||
NM_001198543.1 | NP_001185472.1 | 2, 3 | 215 | |||
NM_001270974.2 | NP_001257903.1 | 2, 3 | 313 | |||
NM_017558.4 | NP_060028.2 | 2, 3 | 313 | |||
XM_006721206.2 | XP_006721269.1 | 2, 3 | 207 | |||
XM_011523146.2 | XP_011521448.1 | 2, 3 | 240 | |||
XM_011523147.1 | XP_011521449.1 | 2, 3 | 207 | |||
XM_011523148.1 | XP_011521450.1 | 2, 3 | 280 | |||
XM_011523151.1 | XP_011521453.1 | 2, 3 | 239 | |||
XM_017023346.1 | XP_016878835.1 | 2, 3 | 485 | |||
GenBank mRNA | AK022933.1 | - | 2, 3 | 247 | ||
AK299016.1 | - | 2, 3 | 202 | |||
AK299348.1 | - | 2, 3 | 245 | |||
AK308296.1 | - | 1, 2 | 32 | |||
AL705531.1 | - | 2, 3 | 257 | |||
BC028351.1 | - | 2, 3 | 194 | |||
BP228881.1 | - | 2, 3 | 232 | |||
DA758908.1 | - | 2, 3 | 280 | |||
DB079205.1 | - | 2, 3 | 280 |