Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286075.1 | NP_001273004.1 | 11 | 1313 | ||
NM_004320.4 | NP_004311.1 | 13 | 1664 | |||
NM_173201.3 | NP_775293.1 | 13 | 1664 | |||
GenBank mRNA | AK128456.1 | - | 11 | 1303 | ||
AK291314.1 | - | 13 | 1593 | |||
BC037354.1 | - | 13 | 1592 | |||
BX537389.1 | - | 13 | 1660 | |||
BX537784.1 | - | 13 | 1662 | |||
BX647365.1 | - | 13 | 1660 | |||
BX647366.1 | - | 13 | 1656 | |||
BX647367.1 | - | 13 | 1664 | |||
BX647368.1 | - | 6 | 702 | |||
BX648544.1 | - | 13 | 1661 | |||
BX649072.1 | - | 13 | 1652 |