Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286075.1 | NP_001273004.1 | 13 | 1882 | ||
NM_004320.4 | NP_004311.1 | 15 | 2233 | |||
NM_173201.3 | NP_775293.1 | 15 | 2233 | |||
GenBank mRNA | AK092691.1 | - | 1 | 30 | ||
AK128456.1 | - | 13 | 1872 | |||
AK291314.1 | - | 15 | 2162 | |||
BC037354.1 | - | 15 | 2161 | |||
BX537389.1 | - | 15 | 2229 | |||
BX537784.1 | - | 15 | 2231 | |||
BX647365.1 | - | 15 | 2229 | |||
BX647366.1 | - | 15 | 2225 | |||
BX647367.1 | - | 15 | 2233 | |||
BX647368.1 | - | 8 | 1271 | |||
BX648544.1 | - | 15 | 2230 | |||
BX649072.1 | - | 15 | 2221 |