Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000507.3 | NP_000498.2 | 6, 7 | 1165 | ||
NM_001127628.1 | NP_001121100.1 | 7, 8 | 1064 | |||
XM_006717005.3 | XP_006717068.1 | 6, 7 | 878 | |||
GenBank mRNA | AF073475.1 | - | 6, 7 | 831 | ||
AK292199.1 | - | 7, 8 | 1103 | |||
AK316583.1 | - | 6, 7 | 1166 | |||
AY544122.1 | - | 6, 7 | 921 | |||
BC012927.1 | - | 6, 7 | 1031 | 4517633 | ||
D26054.1 | - | 6, 7 | 840 | |||
D26055.1 | - | 6, 7 | 840 | |||
D26056.1 | - | 6, 7 | 840 | |||
L10320.1 | - | 6, 7 | 976 | |||
M19922.1 | - | 6, 7 | 1032 |