Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024769.3 | NP_079045.1 | 3 | 576 | ||
XM_011542998.2 | XP_011541300.1 | 3 | 544 | |||
GenBank mRNA | AK026068.1 | - | 3 | 552 | ||
AK304843.1 | - | 3 | 523 | |||
AY326422.1 | - | 3 | 400 | |||
AY358340.1 | - | 3 | 400 | |||
BC009371.2 | - | 3 | 222 | 4122648 | ||
BE272517.1 | - | 3 | 470 | |||
BG723094.1 | - | 3 | 582 | |||
CA395814.1 | - | 3 | 489 |