Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304467.1 | NP_001291396.1 | 3 | 365 | ||
NM_001304468.1 | NP_001291397.1 | 3 | 365 | |||
NM_212554.3 | NP_997719.2 | 3 | 365 | |||
GenBank mRNA | AF318345.1 | - | 3 | 293 | ||
AL832292.1 | - | 3 | 262 | |||
BC026167.1 | - | 3 | 299 | 4823003 | ||
BI551093.1 | - | 3 | 259 | |||
HY048148.1 | - | 3 | 366 |