Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015697.7 | NP_056512.5 | 5 | 883 | ||
XM_011531855.2 | XP_011530157.1 | 5 | 1094 | |||
XM_011531857.2 | XP_011530159.1 | 5 | 1092 | |||
XM_011531859.2 | XP_011530161.1 | 5 | 1091 | |||
XM_011531860.2 | XP_011530162.1 | 5 | 1091 | |||
XM_011531862.2 | XP_011530164.1 | 5 | 1090 | |||
XM_011531863.2 | XP_011530165.1 | 5 | 1087 | |||
XM_011531866.2 | XP_011530168.1 | 5 | 1083 | |||
XM_011531867.2 | XP_011530169.1 | 5 | 1006 | |||
XM_017008031.1 | XP_016863520.1 | 5 | 1006 | |||
XM_017008032.1 | XP_016863521.1 | 3 | 276 | |||
GenBank mRNA | AF091086.1 | - | 4 | 432 | ||
AJ621061.3 | - | 5 | 883 | |||
AK129583.1 | - | 5 | 639 | |||
BC008804.2 | - | 5 | 740 | 3954787 | ||
BC020728.1 | - | 5 | 780 | 4696019 | ||
BQ363759.1 | - | 2 | 159 | |||
CR456860.1 | - | 2 | 142 |