Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127207.1 | NP_001120679.1 | 14 | 2325 | ||
NM_014140.3 | NP_054859.2 | 14 | 2511 | |||
XM_005246631.2 | XP_005246688.1 | 14 | 2449 | |||
XM_005246632.1 | XP_005246689.1 | 14 | 2501 | |||
XM_017004228.1 | XP_016859717.1 | 14 | 2314 | |||
GenBank mRNA | AF082179.1 | - | 13 | 2281 | ||
AF432223.1 | - | 14 | 2321 | |||
AK000117.1 | - | 10 | 1181 | |||
AK023780.1 | - | 14 | 2451 | |||
AL122076.1 | - | 12 | 1745 | |||
BC016482.1 | - | 13 | 2272 | 4341673 | ||
BC029994.1 | - | 11 | 1707 | |||
BC043341.1 | - | 14 | 2298 | 6058575 |