Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UROS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000375.2 | NP_000366.1 | 3 | 343 | ||
NM_001324036.1 | NP_001310965.1 | 3 | 343 | |||
NM_001324037.1 | NP_001310966.1 | 3 | 343 | |||
NM_001324038.1 | NP_001310967.1 | 3 | 343 | |||
NM_001324039.1 | NP_001310968.1 | 3 | 343 | |||
NR_136676.1 | 3 | 343 | ||||
NR_136677.1 | 3 | 343 | ||||
XM_005270140.4 | XP_005270197.1 | 3 | 185 | |||
XM_011540127.1 | XP_011538429.1 | 3 | 185 | |||
XM_017016610.1 | XP_016872099.1 | 3 | 185 | |||
XM_017016611.1 | XP_016872100.1 | 3 | 141 | |||
XM_017016612.1 | XP_016872101.1 | 3 | 185 | |||
GenBank mRNA | AF230665.1 | - | 2 | 210 | ||
AK314896.1 | - | 3 | 200 | |||
BC002573.2 | - | 3 | 302 | 3160463 | ||
J03824.1 | - | 3 | 273 | |||
KU178377.1 | - | 2 | 77 | |||
KU178378.1 | - | 2 | 77 |