Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GJA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000165.4 | NP_000156.1 | 2 | 1295 | ||
GenBank mRNA | AK223289.1 | - | 2 | 1226 | ||
AK294670.1 | - | 3 | 994 | |||
AK295460.1 | - | 3 | 1140 | |||
AK297402.1 | - | 2 | 987 | |||
AK297761.1 | - | 3 | 583 | |||
AK297764.1 | - | 3 | 1179 | |||
AK309377.1 | - | 3 | 691 | |||
AK312324.1 | - | 2 | 1237 | |||
CR541660.1 | - | 1 | 1023 | |||
M65188.1 | - | 2 | 1188 | |||
X52947.1 | - | 2 | 1180 |