Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLDLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018056.2 | NP_001018066.1 | 10 | 1788 | ||
NM_001322225.1 | NP_001309154.1 | 9 | 1665 | |||
NM_001322226.1 | NP_001309155.1 | 9 | 1665 | |||
NM_003383.4 | NP_003374.3 | 10 | 1788 | |||
GenBank mRNA | AB208822.1 | - | 11 | 1544 | ||
AK092381.1 | - | 8 | 1055 | |||
AK095744.1 | - | 6 | 868 | |||
AK289527.1 | - | 2 | 171 | |||
BC136562.1 | - | 10 | 1509 | 9020552 | ||
BC142653.1 | - | 10 | 1450 | 40148037 | ||
BC144245.1 | - | 10 | 1509 | 9052765 | ||
D16493.1 | - | 10 | 1442 | |||
D16494.1 | - | 10 | 1373 | |||
L20470.1 | - | 10 | 1763 | |||
L22431.1 | - | 10 | 1485 | |||
S73849.1 | - | 10 | 1457 |