Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037633.1 | NP_001032722.1 | 5, 6 | 567 | ||
NM_022464.4 | NP_071909.1 | 4, 5 | 479 | |||
XM_011543570.2 | XP_011541872.1 | 6, 7 | 604 | |||
GenBank mRNA | AF547994.1 | - | 4, 5 | 398 | ||
AJ299442.1 | - | 4, 5 | 457 | |||
AK074624.1 | - | 4, 5 | 471 | |||
AK075177.1 | - | 4, 5 | 408 | |||
AY358950.1 | - | 4, 5 | 425 | |||
BC011568.1 | - | 4, 5 | 447 | 4640182 | ||
BI561751.1 | - | 4, 5 | 510 | |||
CB125000.1 | - | 4, 5 | 479 | |||
KU178743.1 | - | 3, 4 | 361 | |||
KU178744.1 | - | 3, 4 | 361 |