Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199199.1 | NP_001186128.1 | 6 | 591 | ||
NM_001319139.1 | NP_001306068.1 | 7 | 733 | |||
NM_001319140.1 | NP_001306069.1 | 7 | 733 | |||
NM_001319141.1 | NP_001306070.1 | 6 | 615 | |||
NM_001319142.1 | NP_001306071.1 | 5 | 489 | |||
NM_001319143.1 | NP_001306072.1 | 5 | 520 | |||
NM_001625.3 | NP_001616.1 | 6 | 615 | |||
NM_013411.4 | NP_037543.1 | 6 | 615 | |||
NR_134976.1 | 5 | 520 | ||||
GenBank mRNA | AB005621.1 | - | 6 | 570 | ||
AB005622.1 | - | 6 | 574 | |||
AB451267.1 | - | 6 | 532 | |||
AB451394.1 | - | 6 | 532 | |||
AK291676.1 | - | 6 | 552 | |||
AK295105.1 | - | 7 | 666 | |||
AK296863.1 | - | 6 | 525 | |||
AK297036.1 | - | 6 | 491 | |||
AK307731.1 | - | 5 | 418 | |||
AL557311.3 | - | 6 | 588 | |||
BC009405.2 | - | 6 | 543 | 4304794 | ||
BC026705.1 | - | 6 | 551 | |||
BC070127.1 | - | 6 | 557 | 6663039 | ||
BC090040.1 | - | 6 | 557 | 6471696 | ||
BQ889289.1 | - | 7 | 699 | |||
DN601721.1 | - | 5 | 439 | |||
U39945.1 | - | 6 | 565 | |||
U54645.1 | - | 6 | 535 | |||
U84371.1 | - | 6 | 563 |